Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

306797

Sigma-Aldrich

(S)-(−)-2-Chloropropionic acid

99%

Synonym(s):

(S)-(−)-2-Chloropropanoic acid, L-α-Chloropropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(Cl)CO2H
CAS Number:
Molecular Weight:
108.52
Beilstein:
1720257
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

99%

form

liquid

optical activity

[α]25/D −14°, neat

refractive index

n20/D 1.4347 (lit.)

bp

77 °C/10 mmHg (lit.)

density

1.249 g/mL at 25 °C (lit.)

SMILES string

C[C@H](Cl)C(O)=O

InChI

1S/C3H5ClO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/t2-/m0/s1

InChI key

GAWAYYRQGQZKCR-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-2-Chloropropionic acid may be used in the preparation of (S)-(-)-2-chloropropionyl chloride[1] and DOTMA ([αR*(αR*,α′R*,α”R*,α”′R*)]-(α,α′,α”,α”′)-tetramethyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid) ligand.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new ytterbium chelate as contrast agent in chemical shift imaging and temperature sensitive probe for MR spectroscopy.
Aime S, et al.
Magnetic Resonance in Medicine : Official Journal of the Society of Magnetic Resonance in Medicine / Society of Magnetic Resonance in Medicine, 35(5), 648-651 (1996)
Synthesis of optically pure. alpha.-alkylated. alpha.-amino acids and a single-step method for enantiomeric excess determination.
Kruizinga WH, et al.
The Journal of Organic Chemistry, 53(8) (1988)
E A Lock et al.
Toxicology, 123(1-2), 41-51 (1997-11-05)
Administration of a single oral dose of 750 mg/kg L-2-chloropropionic acid (L-CPA) to rats produces marked necrosis to the granule cell layer of the cerebellum by 48 h after dosing. Associated with the neuropathology the rats show locomotor impairment and
Gábor Bazsó et al.
The journal of physical chemistry. A, 116(20), 4823-4832 (2012-05-05)
Former assignments of the matrix-isolation infrared (MI-IR) spectrum of 2-chloropropionic acid are revised with the help of near-infrared (NIR) laser irradiation induced change in conformer ratios. This method allows not only the unambiguous assignment of each band in the MI-IR
B B Aam et al.
Toxicology, 228(2-3), 124-134 (2006-09-30)
(+/-)-2-Chloropropionic acid (2-CPA) is a neurotoxic compound which kills cerebellar granule cells in vivo, and makes cerebellar granule cells in vitro produce reactive oxygen species (ROS). We have studied the effect of 2-CPA on ROS formation in human neutrophil granulocytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service