Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

306606

Sigma-Aldrich

2-Fluoro-4-iodoaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(I)NH2
CAS Number:
Molecular Weight:
237.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

55-57 °C (lit.)

SMILES string

Nc1ccc(I)cc1F

InChI

1S/C6H5FIN/c7-5-3-4(8)1-2-6(5)9/h1-3H,9H2

InChI key

CUMTUBVTKOYYOU-UHFFFAOYSA-N

General description

The profiling of iodine-containing metabolites produced by the earthworm Eisenia veneta by exposing it to 2-fluoro-4-iodoaniline was carried out using high-performance liquid chromatography/inductively coupled plasma mass spectrometry (HPLC/ICPMS)[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine J Duckett et al.
Rapid communications in mass spectrometry : RCM, 17(16), 1855-1858 (2003-07-24)
High-performance liquid chromatography/inductively coupled plasma mass spectrometry (HPLC/ICPMS) provided a rapid and specific means for profiling the iodine-containing metabolites produced by the earthworm Eisenia veneta following exposure to 2-fluoro-4-iodoaniline. Profiles were obtained, using gradient reversed-phase HPLC, from extracts of whole

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service