Skip to Content
MilliporeSigma
All Photos(1)

Documents

306088

Sigma-Aldrich

2-Bromo-5-chlorobenzotrifluoride

97%

Synonym(s):

2-Bromo-5-chloro-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)CF3
CAS Number:
Molecular Weight:
259.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.5080 (lit.)

bp

179-180 °C (lit.)

density

1.759 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cc(Cl)ccc1Br

InChI

1S/C7H3BrClF3/c8-6-2-1-4(9)3-5(6)7(10,11)12/h1-3H

InChI key

OSTIALFVJOFNPP-UHFFFAOYSA-N

Application

2-Bromo-5-chlorobenzotrifluoride was used in preparation of 6-[4-chloro-2-(trifluoromethyl)phenyl]-3-fluoro-2-methylpyridine and 4,4′-dichloro-2,2′-bis(trifluoromethyl) diphenyl sulphone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation of a semicrystalline trifluoromethylated poly (aryl ether sulphone).
Clark JH, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 32(6), 1185-1187 (1994)
S Sreenivasa et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 12), o3370-o3370 (2013-03-12)
In the title compound, C13H8ClF4N, the dihedral angle between the benzene and pyridine rings is 59.8 (3)°. In the crystal, mol-ecules are stacked in columns along the b axis through weak C-H⋯π inter-actions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service