Skip to Content
MilliporeSigma
All Photos(1)

Documents

305286

Sigma-Aldrich

1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H26
CAS Number:
Molecular Weight:
446.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

254-256 °C (lit.)

solubility

pyridine: soluble 5 mg/mL, clear, colorless to light yellow

SMILES string

c1ccc(cc1)C2C(c3ccccc3)=C(c4ccccc4)C(c5ccccc5)=C2c6ccccc6

InChI

1S/C35H26/c1-6-16-26(17-7-1)31-32(27-18-8-2-9-19-27)34(29-22-12-4-13-23-29)35(30-24-14-5-15-25-30)33(31)28-20-10-3-11-21-28/h1-25,31H

InChI key

YGLVWOUNCXBPJF-UHFFFAOYSA-N

Application

1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant:
  • for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency
  • in the preparation of monodisperse and highly fluorescent silica nanoparticles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement of electroluminescence efficiency in poly (3-alkylthiophene) by doping of dye molecules.
Ohmori Y, et al.
Synthetic Metals, 85(1), 1241-1242 (1997)
A facile and general approach for the synthesis of fluorescent silica nanoparticles doped with inert dyes.
Mu X, et al.
Mu X, et al.
Chinese Science Bulletin, 56(31), 3242-3246 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service