Skip to Content
MilliporeSigma
All Photos(1)

Documents

304271

Sigma-Aldrich

5-Amino-3-methylisoxazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O
CAS Number:
Molecular Weight:
98.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

85-87 °C (lit.)

SMILES string

Cc1cc(N)on1

InChI

1S/C4H6N2O/c1-3-2-4(5)7-6-3/h2H,5H2,1H3

InChI key

FNXYWHTZDAVRTB-UHFFFAOYSA-N

General description

5-Amino-3-methylisoxazole reacts with α,β-unsaturated ketones to yield the corresponding isoxazolo[5,4-b]pyridines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wafaa S Hamama et al.
Archiv der Pharmazie, 345(6), 468-475 (2012-03-03)
The reaction of 5-amino-3-methylisoxazole with appropriate α,β-unsaturated ketones gave the corresponding isoxazolo[5,4-b]pyridines. Treatment of 1 with 2,6-dibenzylidenecyclohexanone or 2-benzylidenedimedone afforded the corresponding isoxazolo[5,4-b]quinoline derivatives. 4,6,8,9-Tetrahydroisoxazolo[5,4-b]quinolin-5-one derivative was also obtained by multicomponent condensation reaction of 1 with dimedone and benzaldehyde. Heterocyclic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service