Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

303534

Sigma-Aldrich

3-Carboxypyridinium dichromate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12Cr2N2O11
CAS Number:
Molecular Weight:
464.22

Assay

97%

SMILES string

OC(=O)c1cccnc1.OC(=O)c2cccnc2.O[Cr](=O)(=O)O[Cr](O)(=O)=O

InChI

1S/2C6H5NO2.2Cr.2H2O.5O/c2*8-6(9)5-2-1-3-7-4-5;;;;;;;;;/h2*1-4H,(H,8,9);;;2*1H2;;;;;/q;;2*+1;;;;;;;/p-2

InChI key

GWVIYPPLSGDFRR-UHFFFAOYSA-L


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service