Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

302848

Sigma-Aldrich

Pentakis(methoxycarbonyl)cyclopentadiene

97%

Synonym(s):

1,2,3,4,5-Pentacarbomethoxycyclopentadiene, 1,3-Cyclopentadiene-1,2,3,4,5-pentacarboxylic acid, 1,2,3,4,5-pentamethyl ester, Pentamethyl cyclopentadiene-1,2,3,4,5-pentacarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O10
CAS Number:
Molecular Weight:
356.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

149 °C (dec.) (lit.)

functional group

ester

SMILES string

COC(=O)C1C(C(=O)OC)=C(C(=O)OC)C(C(=O)OC)=C1C(=O)OC

InChI

1S/C15H16O10/c1-21-11(16)6-7(12(17)22-2)9(14(19)24-4)10(15(20)25-5)8(6)13(18)23-3/h6H,1-5H3

InChI key

NJNIOTQAGXGMFK-UHFFFAOYSA-N

Application

Useful synthetic building block demonstrated by Tristan Lambert to enable access to chiral Brønsted acid catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service