Skip to Content
MilliporeSigma
All Photos(2)

Documents

301949

Sigma-Aldrich

(1R)-(−)-10-Camphorsulfonyl chloride

97%

Synonym(s):

(−)-Camphor-10-sulfonyl chloride, (1R)-Camphor-10-sulfonic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15ClO3S
CAS Number:
Molecular Weight:
250.74
Beilstein:
3205975
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]18/D −33°, c = 3 in chloroform

mp

66-68 °C (lit.)

SMILES string

[H][C@]12CC[C@](CS(Cl)(=O)=O)(C(=O)C1)C2(C)C

InChI

1S/C10H15ClO3S/c1-9(2)7-3-4-10(9,8(12)5-7)6-15(11,13)14/h7H,3-6H2,1-2H3/t7-,10-/m0/s1

InChI key

BGABKEVTHIJBIW-XVKPBYJWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Jira et al.
Die Pharmazie, 48(11), 829-833 (1993-11-01)
Investigations on direct separation by RP-HPLC of selected enantiomeric beta-adrenergic active agents are described. R- and S-1-phenylethylisocyanate (PEIC) as well as (1S)-(+)-campher-10-sulfonylchloride (CSC) are used for the derivatization of the compounds. Correlations between chromatographic data and various influences (pH, temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service