Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

301809

Sigma-Aldrich

4,7-Dimethyl-1,10-phenanthroline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2
CAS Number:
Molecular Weight:
208.26
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

mp

193-195 °C (lit.)

SMILES string

Cc1ccnc2c1ccc3c(C)ccnc23

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mads Corvinius Nielsen et al.
Bioorganic & medicinal chemistry, 17(24), 8241-8246 (2009-11-10)
A series of 4,7-diamino-1,10-phenanthroline derivatives carrying positively charged side chains has been synthesized, and their G-quadruplex interaction evaluated by circular dichroism (CD) and surface plasmon resonance (SPR). In absence of side chains, 4,7-diamino-1,10-phenanthroline exhibits a weak but significant G-quadruplex stabilizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service