Skip to Content
MilliporeSigma
All Photos(1)

Documents

301639

Sigma-Aldrich

α,α,α,2,3,5,6-Heptafluoro-p-xylene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3F7
CAS Number:
Molecular Weight:
232.10
MDL number:

Assay

98%

SMILES string

Cc1c(F)c(F)c(c(F)c1F)C(F)(F)F

InChI

1S/C8H3F7/c1-2-4(9)6(11)3(8(13,14)15)7(12)5(2)10/h1H3

InChI key

JBZHWNXMZYBXDQ-UHFFFAOYSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service