Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

301612

Sigma-Aldrich

9-Anthraldehyde oxime, predominantly syn

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO
CAS Number:
Molecular Weight:
221.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

162-164 °C (lit.)

functional group

amine
oxime

SMILES string

O\N=C\c1c2ccccc2cc3ccccc13

InChI

1S/C15H11NO/c17-16-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)15/h1-10,17H/b16-10+

InChI key

HLGMTCSSRBPNOQ-MHWRWJLKSA-N

General description

Gallium triflate catalyzed dehydration of 9-anthraldehyde oxime to nitrile is reported.[1]

Application

9-Anthraldehyde oxime was used in preparation of new triosmium cluster [Os3(CO)11(C14H9CN)].[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structural characterization of a triosmium carbonyl cluster containing a 9-anthracenecarbonitrile ligand derived from 9-anthraldehyde oxime.
Wong JS-Y, et al.
Inorganic Chemistry Communications, 6(6), 713-717 (2003)
Gallium (III) triflate catalyzed dehydration of aldoximes.
Yan P, et al.
Catalysis Letters, 101(3), 141-143 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service