Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300748

Sigma-Aldrich

2′,6′-Dimethoxyacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCH3
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

135-136 °C/2 mmHg (lit.)

mp

68-70 °C (lit.)

functional group

ketone

SMILES string

COc1cccc(OC)c1C(C)=O

InChI

1S/C10H12O3/c1-7(11)10-8(12-2)5-4-6-9(10)13-3/h4-6H,1-3H3

InChI key

XEUGKOFTNAYMMX-UHFFFAOYSA-N

General description

2,6-Dihydroxyacetophenone, its mono- and di-methyl ethers are inhibitors of hepatic mixed function oxidases. Metabolism of 2′,6′-dimethoxyacetophenone was studied.[1]

Application

2′,6′-Dimethoxyacetophenone was used in preparation of 4-fluororesorcinol.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile preparations of 4-fluororesorcinol.
Belanger PC, et al.
Canadian Journal of Chemistry, 66(6), 1479-1482 (1988)
A Bobik et al.
Xenobiotica; the fate of foreign compounds in biological systems, 5(2), 65-72 (1975-02-01)
1. 2,6-Dihydroxyacetophenone, its mono- and di-methyl ethers are inhibitors of hepatic mixed function oxidases. The dimethyl ether is a competitive inhibitor of aminopyrine demethylase with the others displaying mixed kinetics. The metabolism of all three ketones has been studied. 2.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service