Skip to Content
MilliporeSigma
All Photos(1)

Documents

300497

Sigma-Aldrich

(2R,3R)-(+)-3-(4-Nitrophenyl)glycidol

98%

Synonym(s):

trans-(+)-2,3-Epoxy-3-(p-nitrophenyl)-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO4
CAS Number:
Molecular Weight:
195.17
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:

Assay

98%

SMILES string

OC[C@H]1O[C@@H]1c2ccc(cc2)[N+]([O-])=O

InChI

1S/C9H9NO4/c11-5-8-9(14-8)6-1-3-7(4-2-6)10(12)13/h1-4,8-9,11H,5H2/t8-,9-/m1/s1

InChI key

TVWSYFXHQPGITR-RKDXNWHRSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service