Skip to Content
MilliporeSigma
All Photos(1)

Documents

300438

Sigma-Aldrich

(2R)-(−)-Glycidyl 4-nitrobenzoate

98%

Synonym(s):

(R)-(−)-Glycidyl 4-nitrobenzoate, (R)-(−)-Oxirane-2-methanol 4-nitrobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO5
CAS Number:
Molecular Weight:
223.18
Beilstein:
5278590
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]19/D −37°, c = 1.08 in chloroform

mp

60-62 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(cc1)C(=O)OC[C@H]2CO2

InChI

1S/C10H9NO5/c12-10(16-6-9-5-15-9)7-1-3-8(4-2-7)11(13)14/h1-4,9H,5-6H2/t9-/m1/s1

InChI key

MUWIANZPEBMVHH-SECBINFHSA-N

Application

(2R)-(−)-Glycidyl 4-nitrobenzoate can be used as a starting material in the total synthesis of leukotriene B4.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of leukotriene B4.
Kerdesky FAJ, et al.
The Journal of Organic Chemistry, 58(13), 3516-3520 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service