Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

300357

Sigma-Aldrich

1,4-Dihydro-2-methylbenzoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O2
CAS Number:
Molecular Weight:
138.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

73-75 °C (lit.)

functional group

carboxylic acid

SMILES string

CC1=CCC=CC1C(O)=O

InChI

1S/C8H10O2/c1-6-4-2-3-5-7(6)8(9)10/h3-5,7H,2H2,1H3,(H,9,10)

InChI key

GYUSHSKDGRZQOB-UHFFFAOYSA-N

General description

Reaction between benzophenone and 1,4-dihydro-2-methylbenzoic acid was studied by combination of steady-state photolysis, laser flash photolysis and photochemically induced dynamic nuclear polarization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inmaculada Andreu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(36), 10089-10096 (2011-07-28)
A thorough mechanistic study has been performed on the reaction between benzophenone (BZP) and a series of 1,4-dienes, including 1,4-cyclohexadiene (CHD), 1,4-dihydro-2-methylbenzoic acid (MBA), 1,4-dihydro-1,2-dimethylbenzoic acid (DMBA) and linoleic acid (LA). A combination of steady-state photolysis, laser flash photolysis (LFP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service