Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300128

Sigma-Aldrich

2-Bromo-9-fluorenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7BrO
CAS Number:
Molecular Weight:
259.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
96%

Assay

96%

form

solid

mp

146-148 °C (lit.)

functional group

bromo
ketone

SMILES string

Brc1ccc2-c3ccccc3C(=O)c2c1

InChI

1S/C13H7BrO/c14-8-5-6-10-9-3-1-2-4-11(9)13(15)12(10)7-8/h1-7H

InChI key

MTCARZDHUIEYMB-UHFFFAOYSA-N

Application

2-Bromo-9-fluorenone was used:
  • as end-capping agent for poly(9,9-dialkylfluorene-2,7-diyl) derivatives
  • in preparation of spirobifluorene ligands
  • in synthesis of altitudinal molecular motors which contain functional groups in their rotor part

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidative stability and its effect on the photoluminescence of poly (fluorene) derivatives: end group effects.
Lee J-I, et al.
Chemistry of Materials, 11(4), 1083-1088 (1999)
Novel iridium complexes as high-efficiency yellow and red phosphorescent light emitters for organic light-emitting diodes.
Yao JH, et al.
Tetrahedron, 64(48), 10814-10820 (2008)
Gábor London et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(32), 10690-10697 (2013-06-21)
We report the synthesis of altitudinal molecular motors that contain functional groups in their rotor part. In an approach to achieve dynamic control over the properties of solid surfaces, a hydrophobic perfluorobutyl chain and a relatively hydrophilic cyano group were
Guang-Wei Zhang et al.
International journal of molecular sciences, 14(11), 22368-22379 (2013-11-16)
Supramolecular luminescence stems from non-covalent exciton behaviors of active π-segments in supramolecular entities or aggregates via intermolecular forces. Herein, a π-conjugated oligofluorenol, containing self-complementary double hydrogen bonds, was synthesized using Suzuki coupling as a supramolecular semiconductor. Terfluorenol-based random supramolecular polymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service