Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

298131

Sigma-Aldrich

Lutetium(III) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Lutetium trichloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LuCl3 · 6H2O
CAS Number:
Molecular Weight:
389.42
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

grade

for analytical purposes

Assay

99.9% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: lutetium

technique(s)

NMR: suitable

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.O.Cl[Lu](Cl)Cl

InChI

1S/3ClH.Lu.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

SJOQNHYDCUXYED-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Employed in synthesis and NMR studies of diamagnetic complexes which exhibit variability of solution structures.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaser H Gholami et al.
International journal of nanomedicine, 15, 31-47 (2020-02-06)
Using our chelate-free, heat-induced radiolabeling (HIR) method, we show that a wide range of metals, including those with radioactive isotopologues used for diagnostic imaging and radionuclide therapy, bind to the Feraheme (FH) nanoparticle (NP), a drug approved for the treatment
Silvio Aime et al.
Inorganic chemistry, 36(14), 2992-3000 (1997-07-02)
The synthesis of three triazamacrocycles containing the pyridine moiety and three acetate pendant arms (PCTA) is reported. The three systems differ due to the number of carbon atoms in the macrocyclic ring forming ligands PCTA-[12], -[13], and -[14], endowed with

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service