Skip to Content
MilliporeSigma
All Photos(1)

Documents

296481

Sigma-Aldrich

5-Chloro-1,3-benzodioxole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClO2
CAS Number:
Molecular Weight:
156.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.556 (lit.)

bp

185-187 °C (lit.)

density

1.34 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc2OCOc2c1

InChI

1S/C7H5ClO2/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3H,4H2

InChI key

ODQPZHOXLYATLC-UHFFFAOYSA-N

General description

5-Chloro-1,3-benzodioxole was converted into a safrole derivative via one-pot process involving two consecutive irradiations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide Ravelli et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(2), 572-579 (2011-01-06)
A mild and general method for the synthesis of potentially bioactive 2-substituted-1,3-benzodioxoles is presented. This is based on the photocatalyzed activation of methylene hydrogen atoms in the presence of tetrabutylammonium decatungstate (TBADT). The method gave yields ranging from 46-77 % with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service