Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

294764

Sigma-Aldrich

tert-Butyldimethylsilane

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$172.00
25 G
$405.00

$172.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$172.00
25 G
$405.00

About This Item

Linear Formula:
(CH3)3CSiH(CH3)2
CAS Number:
Molecular Weight:
116.28
Beilstein:
2231162
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$172.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.4 (lit.)

bp

81-83 °C (lit.)

density

0.701 g/mL at 25 °C (lit.)

SMILES string

C[SiH](C)C(C)(C)C

InChI

1S/C6H16Si/c1-6(2,3)7(4)5/h7H,1-5H3

InChI key

QCIWZIYBBNEPKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raffaele Nicastro et al.
Biochimica et biophysica acta, 1853(7), 1615-1625 (2015-04-07)
The metabolism of proliferating cells shows common features even in evolutionary distant organisms such as mammals and yeasts, for example the requirement for anabolic processes under tight control of signaling pathways. Analysis of the rewiring of metabolism, which occurs following

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service