Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

294462

Sigma-Aldrich

p-Iodo-D-phenylalanine

97%

Synonym(s):

2-Amino-3-(4-iodophenyl)-propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H10NO2I
CAS Number:
Molecular Weight:
291.09
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]25/D −22°, c = 1 in 1 N HCl: ethanol, 1:1

reaction suitability

reaction type: solution phase peptide synthesis

mp

251 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@H](Cc1ccc(I)cc1)C(O)=O

InChI

1S/C9H10INO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m1/s1

InChI key

PZNQZSRPDOEBMS-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service