Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

294152

Sigma-Aldrich

2,3-Diphenylmaleic anhydride

98%

Synonym(s):

3,4-Diphenyl-2,5-furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10O3
CAS Number:
Molecular Weight:
250.25
Beilstein:
191180
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

mp

159-162 °C (lit.)

SMILES string

O=C1OC(=O)C(c2ccccc2)=C1c3ccccc3

InChI

1S/C16H10O3/c17-15-13(11-7-3-1-4-8-11)14(16(18)19-15)12-9-5-2-6-10-12/h1-10H

InChI key

OUJCFCNZIUTYBH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pawlowska et al.
Journal of chromatography. A, 666(1-2), 485-491 (1994-04-22)
New pre-column derivatizing reagents: phthalic anhydride, 3-nitrophthalic anhydride, diphenic anhydride, 1,8-naphthalic anhydride and diphenylmaleic anhydride have been developed for resolving chiral compounds having amine groups. Although all of these agents produce derivatives with high molar absorptivities, the later two also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service