Skip to Content
MilliporeSigma
All Photos(1)

Documents

293792

Sigma-Aldrich

Methylthiomethyl p-tolyl sulfone

98%

Synonym(s):

Methylmercaptomethyl p-tolyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2CH2SCH3
CAS Number:
Molecular Weight:
216.32
Beilstein:
2259685
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

82-85 °C (lit.)

SMILES string

CSCS(=O)(=O)c1ccc(C)cc1

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service