Skip to Content
MilliporeSigma
All Photos(1)

Documents

293687

Sigma-Aldrich

Dibenzyl diselenide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2Se2
CAS Number:
Molecular Weight:
340.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

91-93 °C (lit.)

SMILES string

C([Se][Se]Cc1ccccc1)c2ccccc2

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yapei Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 22(13), 5552-5555 (2006-06-14)
We report on the use of block copolymer micelles of polystyrene-b-poly(acrylic acid) (PS-b-PAA) as matrixes for incorporating dibenzyl diselenide. We found that the water-insoluble diselenide, after being incorporated into the micelles, demonstrates glutathione peroxidase (GPx) activity in water. Surprisingly, the
T Shimada et al.
Cancer research, 57(21), 4757-4764 (1997-11-14)
The effects of synthetic chemopreventive organoselenium compounds 1,2-, 1,3-, and 1,4-phenylenebis(methylene)selenocyanate (o-, m-, and p-XSC, respectively), benzyl selenocyanate (BSC), and dibenzyl diselenide (DDS) and inorganic sodium selenite on the oxidation of xenobiotics and procarcinogens by human cytochrome P450 (P450 or
K el-Bayoumy et al.
Chemical research in toxicology, 4(5), 560-565 (1991-09-01)
Benzyl selenocyanate (BSC), a synthetic organoselenium compound, has been shown to inhibit chemically induced tumors in several animal model systems. However, it is not known whether BSC or one of its metabolites is responsible for the chemopreventive effect. An initial
B S Reddy et al.
Anticancer research, 16(3A), 1123-1127 (1996-05-01)
Our previous studies have demonstrated that dietary benzylselenocyanate (BSC) and 1,4-phenylenebis (methylene) selenocyanate (p-XSC); organoselenium compounds, act as potential chemopreventive agents in colon carcinogenesis in F344 rats. As a part of a program aimed to develop less toxic and more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service