Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

292923

Sigma-Aldrich

DL-Lactamide

97%

Synonym(s):

DL-Lactic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CONH2
CAS Number:
Molecular Weight:
89.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

73-76 °C (lit.)

solubility

water: soluble 25 mg/mL, clear to slightly hazy, colorless

SMILES string

CC(O)C(N)=O

InChI

1S/C3H7NO2/c1-2(5)3(4)6/h2,5H,1H3,(H2,4,6)

InChI key

SXQFCVDSOLSHOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Lactamide is a hyperosmotic agent and its effect on ciliary epithelium and trabecular meshwork of Rhesus monkey has been investigated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Mack et al.
ChemSusChem, 12(8), 1653-1663 (2019-02-16)
Acrylonitrile (ACN) is widely used as monomer in the synthesis of polymers and carbon fibers. Nowadays, its production is based on fossil resources. Herein, an alternative process based on renewable resources is presented. Lactic acid (LA), which can be obtained
S Okisaka et al.
Investigative ophthalmology, 15(8), 617-625 (1976-08-01)
Hyperosmotic agents (2 M urea or 2 M DL-lactamide), perfused into the internal carotid artery of Rhesus monkeys, produced a marked decrease in intraocular pressure and an increase in aqueous humor protein. Fenestrae of the ciliary capillaries were broken immediately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service