Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

292850

Sigma-Aldrich

Bis(2-ethylhexyl)amine

99%

Synonym(s):

Diisooctylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CH2]2NH
CAS Number:
Molecular Weight:
241.46
Beilstein:
1748342
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.443 (lit.)

bp

123 °C/5 mmHg (lit.)

density

0.805 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCC(CC)CNCC(CC)CCCC

InChI

1S/C16H35N/c1-5-9-11-15(7-3)13-17-14-16(8-4)12-10-6-2/h15-17H,5-14H2,1-4H3

InChI key

SAIKULLUBZKPDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-ethylhexyl)amine constitutes surfactant-based liquid mixtures having potential specialized applications[1].

Application

Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules[2].

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

266.0 °F - Pensky-Martens closed cup

Flash Point(C)

130 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Calandra et al.
Journal of colloid and interface science, 348(1), 183-188 (2010-05-15)
Surfactant-based liquid mixtures constitute an interesting class of nanostructured materials with promising potential in specialized applications. Here, structural and conductometric properties of liquid mixtures composed of bis(2-ethylhexyl)amine (BEEA) and bis(2-ethylhexyl)phosphoric acid (HDEHP) have been thoroughly investigated with the aim to
Synthesis, size control, and passivation of CdS nanoparticles in water/AOT/ n-heptane microemulsions.
Caponetti E, et al.
Materials Science & Engineering. C, Materials For Biological Applications, 23(4), 531-539 (2003)
Clayton J Dahlman et al.
ACS nano, 14(9), 11294-11308 (2020-08-25)
Controlling the structure of layered hybrid metal halide perovskites, such as the Ruddlesden-Popper (R-P) phases, is challenging because of their tendency to form mixtures of varying composition. Colloidal growth techniques, such as antisolvent precipitation, form dispersions with properties that match

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service