Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

292346

Sigma-Aldrich

Ethyl 3-(trimethylsilyl)propionate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18O2Si
CAS Number:
Molecular Weight:
174.31
Beilstein:
1759717
MDL number:
UNSPSC Code:
12352100

Assay

95%

density

0.878 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)CC[Si](C)(C)C

InChI

1S/C8H18O2Si/c1-5-10-8(9)6-7-11(2,3)4/h5-7H2,1-4H3

InChI key

JCANPYOQOFSGFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service