Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

292311

Sigma-Aldrich

2,3,6-Trichlorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2CHO
CAS Number:
Molecular Weight:
209.46
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

87-89 °C (lit.)

functional group

aldehyde
chloro

SMILES string

Clc1ccc(Cl)c(C=O)c1Cl

InChI

1S/C7H3Cl3O/c8-5-1-2-6(9)7(10)4(5)3-11/h1-3H

InChI key

AURSMWWOMOVHBM-UHFFFAOYSA-N

General description

The influence of paramagnetic ions, Cu2+ and Mn2+, on the relaxation behavior of long-lived coherences has been studied in a two-spin system, 2,3,6-trichlorobenzaldehyde[1].

Application

2,3,6-Trichlorobenzaldehyde has been used in the preparation of:
  • 2,3,6-trichloro-5-nitrobenzaldehyde[2]
  • 2,3,6-trichloromandelic acid[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maninder Singh et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 14(17), 3977-3981 (2013-10-24)
Long-lived coherences (LLCs) are known to have lifetimes much longer than transverse magnetization or single quantum coherences (SQCs). The effect of paramagnetic ions on the relaxation of LLCs is not known. This is particularly important, as LLCs have potential applications
Hapten synthesis and antibody development for polychlorinated dibenzo-p-dioxin immunoassays.
Sanborn JR, et al.
Journal of Agricultural and Food Chemistry, 46(6), 2407-2416 (1998)
M Keith Davis et al.
Journal of exposure science & environmental epidemiology, 18(6), 564-570 (2008-01-03)
Fleas are a persistent problem for pets that require implementation of control measures. Consequently, pesticide use by homeowners for flea control is common and may increase pesticide exposure for adults and children. Fifty-five pet dogs (23 in study 1; 22

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service