Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

290971

Sigma-Aldrich

2,2-Dimethyl-4-pentenal

technical grade, 90%, contains 1000 ppm hydroquinone as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C(CH3)2CHO
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

contains

1000 ppm hydroquinone as stabilizer

refractive index

n20/D 1.4203 (lit.)

bp

124-125 °C (lit.)

density

0.825 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(CC=C)C=O

InChI

1S/C7H12O/c1-4-5-7(2,3)6-8/h4,6H,1,5H2,2-3H3

InChI key

DXSDIWHOOOBQTJ-UHFFFAOYSA-N

Application

2,2-Dimethyl-4-pentenal has been used:
  • as electrophile in the synthesis of rearranged bicyclo[6.3.0]undecane isoprenoid skeleton of cyclooctenoid sesquiterpene dactylol and 3a-epi-dactylol
  • in the preparation of imine ligand via condensation with primary amine

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Latent olefin metathesis catalysts featuring chelating alkylidenes.
Hejl A, et al.
Organometallics, 25(26), 6149-6154 (2006)
Alois Fürstner et al.
The Journal of organic chemistry, 61(25), 8746-8749 (1996-12-13)
A straightforward total synthesis of the cyclooctenoid sesquiterpene dactylol (1) and of 3a-epi-dactylol (13) has been achieved in six synthetic operations. The unusual rearranged bicyclo[6.3.0]undecane isoprenoid skeleton of these target molecules has been formed via an initial three-component coupling triggered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service