Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

290874

Sigma-Aldrich

N-Methylpropionamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CONHCH3
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.436 (lit.)

bp

146 °C/90 mmHg (lit.)

mp

−43 °C (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)NC

InChI

1S/C4H9NO/c1-3-4(6)5-2/h3H2,1-2H3,(H,5,6)

InChI key

QJQAMHYHNCADNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Methylpropionamide has been used to investigate amide-induced phase separation of hexafluoro-2-propanol-water mixtures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Toshiyuki Takamuku et al.
Physical chemistry chemical physics : PCCP, 14(23), 8335-8347 (2012-05-17)
Amide-induced phase separation of hexafluoro-2-propanol (HFIP)-water mixtures has been investigated to elucidate solvation properties of the mixtures by means of small-angle neutron scattering (SANS), (1)H and (13)C NMR, and molecular dynamics (MD) simulation. The amides included N-methylformamide (NMF), N-methylacetamide (NMA)
Yusuke Asakura et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(7), 1893-1900 (2014-01-17)
AST-type zeolite with a plate morphology can be synthesized by topotactic conversion of a layered silicate (β-helix-layered silicate; HLS) by using N,N-dimethylpropionamide (DPA) to control the layer stacking of silicate layers and the subsequent interlayer condensation. Treatment of HLS twice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service