Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

290645

Sigma-Aldrich

trans-1,2-Dimethylcyclohexane

99%

Synonym(s):

trans-Hexahydro-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
3193642
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

36.6 mmHg ( 37.7 °C)

Assay

99%

autoignition temp.

579 °F

refractive index

n20/D 1.427 (lit.)

bp

123-124 °C (lit.)

mp

−89 °C (lit.)

density

0.77 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CCCC[C@H]1C

InChI

1S/C8H16/c1-7-5-3-4-6-8(7)2/h7-8H,3-6H2,1-2H3/t7-,8-/m1/s1

InChI key

KVZJLSYJROEPSQ-HTQZYQBOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adsorption of trans-1,2-dimethylcyclohexane on chiral single-walled carbon nanotubes has been examined.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy D Power et al.
Journal of the American Chemical Society, 124(9), 1858-1859 (2002-02-28)
Single-walled carbon nanotubes can exist in chiral forms and can adsorb a range of molecules. We use atomistic simulations to consider whether enantiopure carbon nanotubes might be effective enantiospecific adsorbents for chiral molecules. We examine the adsorption of both enantiomers
Frank-Dieter Kopinke et al.
Environmental science & technology, 45(23), 10013-10019 (2011-11-01)
This study presents a new experimental technique for measuring rates of desorption of organic compounds from dissolved organic matter (DOM) such as humic substances. The method is based on a fast solid-phase extraction of the freely dissolved fraction of a
Acute toxicities to larval rainbow trout of representative compounds detected in Great Lakes fish.
C C Edsall
Bulletin of environmental contamination and toxicology, 46(2), 173-178 (1991-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service