Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

290440

Sigma-Aldrich

3,5-Difluorophenylacetic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2CO2H
CAS Number:
Molecular Weight:
172.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

68-70 °C (lit.)

SMILES string

OC(=O)Cc1cc(F)cc(F)c1

InChI

1S/C8H6F2O2/c9-6-1-5(3-8(11)12)2-7(10)4-6/h1-2,4H,3H2,(H,11,12)

InChI key

IGGNSAVLXJKCNH-UHFFFAOYSA-N

General description

3,5-Difluorophenylacetic acid is an important pharmaceutical intermediate.

Application

3,5-Difluorophenylacetic acid has been used in the synthesis of:
  • N-[N-(3,5-difluorophenylacetyl)-L-alanyl]-L-phenylglycine tert-butyl ester
  • N-acylalanine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilles Quéléver et al.
Organic & biomolecular chemistry, 3(13), 2450-2457 (2005-06-25)
Inhibition of gamma-secretase, one of the enzymes responsible for the cleavage of the amyloid precursor protein (APP) to produce pathogenic Abeta peptides, is an attractive approach for the treatment of Alzheimer's disease. We designed a gamma-secretase inhibitor bearing an ascorbic
Synthesis of a tetrahydroimidazo [2', 1': 2, 3] thiazolo [5, 4-c] pyridine derivative with Met inhibitory activity.
Amat M, et al.
ARKIVOC (Gainesville, FL, United States), 3, 145-151 (2010)
Synthesis of dihalophenylacetic acids using aromatic nucleophilic substitution strategy.
Kowalczyk BA.
Synthesis, 1997(12), 1411-1414 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service