Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

290386

Sigma-Aldrich

trans-3,5-Difluorocinnamic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
184.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

204-205 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

[H]\C(=C(\[H])c1cc(F)cc(F)c1)C(O)=O

InChI

1S/C9H6F2O2/c10-7-3-6(1-2-9(12)13)4-8(11)5-7/h1-5H,(H,12,13)/b2-1+

InChI key

MBAWRXICVNIUGY-OWOJBTEDSA-N

Application

3,5-Difluorocinnamic acid has been used in the synthesis of “unnatural” flavonoids and stilbenes in Escherichia coli.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yohei Katsuyama et al.
Chemistry & biology, 14(6), 613-621 (2007-06-23)
Flavonoids and stilbenes have attracted much attention as potential targets for nutraceuticals, cosmetics, and pharmaceuticals. We have developed a system for producing "unnatural" flavonoids and stilbenes in Escherichia coli. The artificial biosynthetic pathway included three steps. These included a substrate
Noah D Bronstein et al.
ACS nano, 13(4), 3839-3846 (2019-03-12)
We present a combined experimental and theoretical study of ligand-ligand cooperativity during X-type carboxylate-to-carboxylate ligand exchange reactions on PbS quantum dot surfaces. We find that the ligand dipole moment (varied through changing the substituents on the benzene ring of cinnamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service