Skip to Content
MilliporeSigma
All Photos(1)

Documents

289965

Sigma-Aldrich

Protoporphyrin IX

Synonym(s):

3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionic acid, Kammerer’s porphyrin, Ooporphyrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H34N4O4
CAS Number:
Molecular Weight:
562.66
Beilstein:
380795
EC Number:
MDL number:
UNSPSC Code:
12352103

SMILES string

Cc1c(CCC(O)=O)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(C)c5C=C)c(C)c4C=C)c(C)c3CCC(O)=O

InChI

1S/C34H34N4O4/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25/h7-8,13-16,35,38H,1-2,9-12H2,3-6H3,(H,39,40)(H,41,42)/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-

InChI key

KSFOVUSSGSKXFI-UJJXFSCMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Activates soluble guanylyl cyclase.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service