Skip to Content
MilliporeSigma
All Photos(1)

Documents

289493

Sigma-Aldrich

N-(2-Chlorobenzyloxycarbonyloxy)succinimide

98%

Synonym(s):

2-Chlorobenzyl succinimidyl carbonate, Z(2-Cl)-OSu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10ClNO5
CAS Number:
Molecular Weight:
283.66
Beilstein:
3623238
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

102-105 °C (lit.)

SMILES string

Clc1ccccc1COC(=O)ON2C(=O)CCC2=O

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service