Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

288489

Sigma-Aldrich

1,3-Dibenzyloxy-2-propanol

97%

Synonym(s):

1,3-Bis(benzyloxy)-2-propanol, 1,3-Di-O-benzylglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2OCH2)2CHOH
CAS Number:
Molecular Weight:
272.34
Beilstein:
1986774
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.549 (lit.)

bp

226-227 °C/3 mmHg (lit.)

density

1.103 g/mL at 20 °C (lit.)

functional group

ether
hydroxyl
phenyl

SMILES string

OC(COCc1ccccc1)COCc2ccccc2

InChI

1S/C17H20O3/c18-17(13-19-11-15-7-3-1-4-8-15)14-20-12-16-9-5-2-6-10-16/h1-10,17-18H,11-14H2

InChI key

ARLSYSVVBAMYKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Dibenzyloxy-2-propanol was used in the synthesis of 1,3-dibenzyloxyacetone[1]. It was also used as precursor to antiviral intermediates[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A trihydroxy acyclonucleoside series.
Ogilvie KK, et al.
Canadian Journal of Chemistry, 62 (8), 1622-1627 (1984)
J C Martin et al.
Journal of medicinal chemistry, 26(5), 759-761 (1983-05-01)
The synthesis of a new acyclic analogue of deoxyguanosine, 9-[(1,3-dihydroxy-2-propoxy)methyl]guanine (DHPG, 1), is described starting from epichlorohydrin via condensation of 2-O-(acetoxymethyl)-1,3-di-O-benzylglycerol (5) with N2,9-diacetylguanine (6). In vitro studies indicate that DHPG is a potent and broad-acting (herpes simplex virus types

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service