Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

288403

Sigma-Aldrich

trans-Dichlorobis(triphenylphosphine)platinum(II)

Synonym(s):

trans-Bis(triphenylphosphine)platinum dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$322.00

$322.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$322.00

About This Item

Linear Formula:
[(C6H5)3P]2PtCl2
CAS Number:
Molecular Weight:
790.56
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$322.00


Please contact Customer Service for Availability

Request a Bulk Order

reaction suitability

core: platinum
reagent type: catalyst

SMILES string

Cl[Pt]Cl.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.2ClH.Pt/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

InChI key

XAFJSPPHVXDRIE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Preparation of soluble polymers of monosubstituted acetylenes with quaternary ammonium pendant groups
  • Alkynylation and hydrogenation reactions
  • Hetero-Diels-Alder/ring-opening reactions
  • Insertion of carbenes into O-H bonds of alcohols
  • Cyclopropanation of olefins
  • Hydroformylation reactions

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system,Peripheral nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service