Skip to Content
MilliporeSigma
All Photos(1)

Documents

287989

Sigma-Aldrich

1,6-Bis(diphenylphosphino)hexane

97%

Synonym(s):

1,6-Hexanediylbis[diphenylphosphine, Hexamethylenebis[diphenylphosphine]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2P(CH2)6P(C6H5)2
CAS Number:
Molecular Weight:
454.52
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

124-126 °C (lit.)

functional group

phosphine

SMILES string

C(CCCP(c1ccccc1)c2ccccc2)CCP(c3ccccc3)c4ccccc4

InChI

1S/C30H32P2/c1(15-25-31(27-17-7-3-8-18-27)28-19-9-4-10-20-28)2-16-26-32(29-21-11-5-12-22-29)30-23-13-6-14-24-30/h3-14,17-24H,1-2,15-16,25-26H2

InChI key

GPORFKPYXATYNX-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service