Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

287377

Sigma-Aldrich

D-Turanose

98%

Synonym(s):

3-O-α-D-Glucopyranosyl-D-fructose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93771
EC Number:
MDL number:
UNSPSC Code:
12352201

Assay

98%

optical activity

[α]20/D +75°, c = 4 in H2O

mp

170 °C (dec.) (lit.)

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@H](O)COC2(O)CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-5-7(17)8(18)9(19)11(22-5)23-10-6(16)4(15)2-21-12(10,20)3-14/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12?/m1/s1

InChI key

SEWFWJUQVJHATO-PUVWEJBASA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Gonzali et al.
The Plant journal : for cell and molecular biology, 44(4), 633-645 (2005-11-03)
Sugars acting as signalling molecules regulate many developmental processes in plants, including lateral and adventitious root production. Turanose, a non-metabolizable sucrose analogue, profoundly affects the growth pattern of Arabidopsis seedlings. Turanose-treated seedlings are characterized by a very short primary root
Alok K Sinha et al.
Plant physiology, 128(4), 1480-1489 (2002-04-16)
To gain insight into the regulatory mechanisms of sugar signaling in plants, the effect of derivatives of the transport sugar sucrose (Suc), the Suc isomers palatinose and turanose, and the Suc analog fluoro-Suc were tested. Photo-autotrophic suspension culture cells of
Rui C Martins et al.
Natural product research, 22(17), 1560-1582 (2008-11-22)
The main purpose of this study was the characterisation of 'Serra da Lousã' heather honey by using novel statistical methodology, relevant principal component analysis, in order to assess the correlations between production year, locality and composition. Herein, we also report
G Potocki de Montalk et al.
FEBS letters, 471(2-3), 219-223 (2000-04-18)
Amylosucrase is a glucosyltransferase that synthesises an insoluble alpha-glucan from sucrose. The catalytic properties of the highly purified amylosucrase from Neisseria polysaccharea were characterised. Contrary to previously published results, it was demonstrated that in the presence of sucrose alone, several
J Defaye et al.
Carbohydrate research, 251, 1-15 (1994-01-03)
Dispirodioxanyl pseudotetrasaccharides 6-O-alpha-D-glucopyranosyl-alpha-D-fructofuranose 6-O-alpha-D-glucopyranosyl-beta-D-fructofuranose 1,2':2,1'-dianhydride, 5-O-alpha-D-glucopyranosyl-alpha-D-fructopyranose 5-O-alpha-D-glucopyranosyl-beta-D-fructopyranose 1,2':2,1'-dianhydride, 4-O-alpha-D-glucopyranosyl-alpha-D-fructofuranose 4-O-alpha-D-glucopyranosyl-beta-D-fructopyranose 1,2':2,1'-dianhydride, 4-O-beta-D-galactopyranosyl-alpha-D-fructofuranose 4-O-beta-D-galactopyranosyl-beta-D-fructopyranose 1,2':2,1'-dianhydride, and 3-O-alpha-D-glucopyranosyl-alpha-D-fructofuranose 3-O-alpha-D-glucopyranosyl-beta-D-fructofuranose 1,2':2,1'-dianhydride were respectively obtained, on a preparative scale, by dissolution of the isomeric glycosylfructoses palatinose, leucrose, maltulose, lactulose, and turanose in anhydrous hydrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service