Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28690

Sigma-Aldrich

Diethyl cyclobutane-1,1-dicarboxylate

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4
CAS Number:
Molecular Weight:
200.23
Beilstein:
2050195
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

bp

104-105 °C/12 mmHg (lit.)

density

1.05 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)C1(CCC1)C(=O)OCC

InChI

1S/C10H16O4/c1-3-13-8(11)10(6-5-7-10)9(12)14-4-2/h3-7H2,1-2H3

InChI key

JPNJEJSZSMXWSV-UHFFFAOYSA-N

Application

Diethyl cyclobutane-1,1-dicarboxylate may be used in chemical synthesis studies.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service