Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

285838

Sigma-Aldrich

Ethopropazine hydrochloride

98%

Synonym(s):

10-(2-Diethylaminopropyl)phenothiazine, 10-[2-(Diethylamino)propyl]phenothiazine hydrochloride, Dibutil hydrochloride, Parkin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2S · HCl
CAS Number:
Molecular Weight:
348.93
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

SMILES string

Cl[H].CCN(CC)C(C)CN1c2ccccc2Sc3ccccc13

InChI

1S/C19H24N2S.ClH/c1-4-20(5-2)15(3)14-21-16-10-6-8-12-18(16)22-19-13-9-7-11-17(19)21;/h6-13,15H,4-5,14H2,1-3H3;1H

InChI key

VXPCQISYVPFYRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ethoproprazine hydrochloride is an inhibitor of butyrylcholinesterase; antiparkinsonian. It reduces extrapyramidal motor effects, characteristic of Parkinson′s disease; also alleviates thermal hyperalgesia in rats.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamil Musilek et al.
European journal of medicinal chemistry, 46(2), 811-818 (2011-01-18)
Inhibitors of acetylcholinesterase are compounds widely used in the treatment of various diseases, such as Alzheimer's disease, glaucoma and Myasthenia gravis (MG). Compounds used in the therapy of MG posses a positive charge in the molecule to ensure peripheral effect
Kamil Musilek et al.
Bioorganic & medicinal chemistry letters, 21(1), 150-154 (2010-12-15)
Carbamate inhibitors (e.g., pyridostimine bromide) are used as a pre-exposure treatment for the prevention of organophosphorus poisoning. They work by blocking acetylcholinesterase's (AChE) native function and thus protect AChE against irreversible inhibition by organophosphorus compounds. However, carbamate inhibitors are known
Marketa Komloova et al.
Bioorganic & medicinal chemistry letters, 21(8), 2505-2509 (2011-03-15)
This paper describes the preparation and in vitro evaluation of 18 newly prepared bis-quinolinium inhibitors on human recombinant acetylcholinesterase (AChE) and human plasmatic butyrylcholinesterase (BChE). Their inhibitory (IC(50)) and was compared to the chosen standards ambenonium dichloride, edrophonium chloride, BW284c51

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service