Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28452

Sigma-Aldrich

Isopropyl cyanoacetate

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2COOCH(CH3)2
CAS Number:
Molecular Weight:
127.14
Beilstein:
1756014
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

refractive index

n20/D 1.416

bp

207 °C (lit.)

density

1.013 g/mL at 20 °C (lit.)

SMILES string

CC(C)OC(=O)CC#N

InChI

1S/C6H9NO2/c1-5(2)9-6(8)3-4-7/h5H,3H2,1-2H3

InChI key

BESQLCCRQYTQQI-UHFFFAOYSA-N

Application

Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing Knöevenagel reactions with aldehydes and ketones[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of isopropyl alcohol as a solvent in Ti (O-i-Pr)4 catalyzed Knoevenagel reactions.
Tetrahedron, 61(33), 7981-7985 (2005)

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service