Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

284238

Sigma-Aldrich

1,2,3-Heptanetriol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH(OH)CH2OH
CAS Number:
Molecular Weight:
148.20
Beilstein:
1737083
MDL number:
UNSPSC Code:
12162002

Assay

98%

mp

52-55 °C (lit.)

SMILES string

CCCCC(O)C(O)CO

InChI

1S/C7H16O3/c1-2-3-4-6(9)7(10)5-8/h6-10H,2-5H2,1H3

InChI key

HXYCHJFUBNTKQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service