Skip to Content
MilliporeSigma
All Photos(1)

Documents

284068

Sigma-Aldrich

Sulforhodamine 101 acid chloride

Dye content 65 %

Synonym(s):

Sulforhodamine 101 sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H29ClN2O6S2
CAS Number:
Molecular Weight:
625.15
Beilstein:
8667894
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 65%

impurities

≤15% CHCl3

λmax

369 nm
587 nm (2nd)

SMILES string

[O-]S(=O)(=O)c1cc(ccc1C2=C3C=C4CCC[N+]5=C4C(CCC5)=C3Oc6c7CCCN8CCCc(cc26)c78)S(Cl)(=O)=O

InChI

1S/C31H29ClN2O6S2/c32-41(35,36)20-9-10-21(26(17-20)42(37,38)39)27-24-15-18-5-1-11-33-13-3-7-22(28(18)33)30(24)40-31-23-8-4-14-34-12-2-6-19(29(23)34)16-25(27)31/h9-10,15-17H,1-8,11-14H2

InChI key

MPLHNVLQVRSVEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service