Skip to Content
MilliporeSigma
All Photos(2)

Documents

283649

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H,23H-porphine vanadium(IV) oxide

Synonym(s):

Oxo[5,10,15,20-tetraphenylporphinato(2-)]vanadium, Vanadyl(IV) meso-tetraphenylporphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H28N4OV
CAS Number:
Molecular Weight:
679.66
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

reaction suitability

reagent type: catalyst
core: vanadium

λmax

423 nm

SMILES string

O=[V]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28N4.O.V/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;;/q-2;;+2/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;

InChI key

WDCQRRQLLCXEFB-YKKPBKTHSA-N

Application

Thin films of VOTPP may be used in detecting methane gas.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thin films for microsensors
Yahaya M and Salleh MM et al.
Advanced Mat. null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service