Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283576

Sigma-Aldrich

2-Ethoxybenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CN
CAS Number:
Molecular Weight:
147.17
Beilstein:
2803453
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
EC Index Number:
613-895-3
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.5322 (lit.)

density

1.053 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccccc1C#N

InChI

1S/C9H9NO/c1-2-11-9-6-4-3-5-8(9)7-10/h3-6H,2H2,1H3

InChI key

DXTLCLWOCYLDHL-UHFFFAOYSA-N

Application

2-Ethoxybenzonitrile has been used in the synthesis of selective phosphodiesterase(V) inhibitor, vardenafil[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of a phosphodiesterase (V) inhibitor Vardenafil [J].
CHEN L, et al.
Chemical Reagents, 5, 011-011 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service