Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283517

Sigma-Aldrich

Ethyl 3-fluorobenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CO2C2H5
CAS Number:
Molecular Weight:
168.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.485 (lit.)

bp

94-95 °C/16 mmHg (lit.)

density

1.136 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1cccc(F)c1

InChI

1S/C9H9FO2/c1-2-12-9(11)7-4-3-5-8(10)6-7/h3-6H,2H2,1H3

InChI key

SMMIKBXLEWTSJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 3-fluorobenzoate has been used in the synthesis of [2-(3-fluorophenyl)allyl]trimethylsilane.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimisation of the allylsilane approach to C-10 deoxo carba analogues of dihydroartemisinin: synthesis and in vitro antimalarial activity of new, metabolically stable C-10 analogues.
O'Neill PM, et al.
Journal of the Chemical Society. Perkin Transactions 1, 20, 2682-2689 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service