Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283495

Sigma-Aldrich

5-Bromo-2-fluorotoluene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Br)F
CAS Number:
Molecular Weight:
189.02
Beilstein:
2242693
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.529 (lit.)

density

1.486 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Cc1cc(Br)ccc1F

InChI

1S/C7H6BrF/c1-5-4-6(8)2-3-7(5)9/h2-4H,1H3

InChI key

VXKYOKPNAXNAFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-2-fluorotoluene has been used in the preparation of:
  • 5-bromo-2-fluorobenzyl bromide via N-bromosuccinimide (NBS) bromination, required for the synthesis of crown bromofluorobenzene
  • 3-(4-fuoro-3-methylphenyl)acrylamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facilitated catecholamine transport through bulk and polymer-supported liquid membranes.
Paugam M-F, et al.
Journal of the American Chemical Society, 118(41), 9820-9825 (1996)
Synthesis of 3-arylpropylamine derivatives from aryl halides using Heck reaction.
Baek GH, et al.
Bull. Korean Chem. Soc., 20(2), 233-233 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service