283177
3-Fluorobenzal bromide
97%
Synonym(s):
1-(Dibromomethyl)-3-fluorobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
FC6H4CHBr2
CAS Number:
Molecular Weight:
267.92
MDL number:
UNSPSC Code:
12352100
Recommended Products
Assay
97%
refractive index
n20/D 1.587 (lit.)
bp
108-110 °C/8 mmHg (lit.)
density
1.915 g/mL at 25 °C (lit.)
SMILES string
Fc1cccc(c1)C(Br)Br
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service