Skip to Content
MilliporeSigma
All Photos(1)

Documents

282529

Sigma-Aldrich

4-Chloro-L-phenylalanine

98%

Synonym(s):

L-PCPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
199.63
Beilstein:
2416150
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

optical activity

[α]20/D -25°, c = 0.5% in H2O

mp

263 °C (dec.) (lit.)

SMILES string

N[C@@H](Cc1ccc(Cl)cc1)C(O)=O

InChI

1S/C9H10ClNO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m0/s1

InChI key

NIGWMJHCCYYCSF-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Tryptophan hydroxylase inhibitor.

replaced by

Product No.
Description
Pricing

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service