Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

282154

Sigma-Aldrich

3-(2-Thienyl)-L-alanine

98%

Synonym(s):

(S)-α-Amino-2-thiophenepropionic acid, (S)-2-Amino-3-(2-thienyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO2S
CAS Number:
Molecular Weight:
171.22
Beilstein:
82872
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −29.4°, c = 1 in H2O

mp

255-263 °C (dec.) (lit.)

SMILES string

N[C@@H](Cc1cccs1)C(O)=O

InChI

1S/C7H9NO2S/c8-6(7(9)10)4-5-2-1-3-11-5/h1-3,6H,4,8H2,(H,9,10)/t6-/m0/s1

InChI key

WTOFYLAWDLQMBZ-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Unusual α-amino acid.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service